ATC311763652 Atlantic Research Chemicals Ltd

(1-Methyl-1H-benzimidazol-2-yl)methylamine

  • MDL number MFCD00457746
  • Cas Number : 20028-40-4
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM02201
Availability details for
Quantity
  • Availability
  • Add to Cart