ATC311763682 Atlantic Research Chemicals Ltd

2-[(Dimethylamino)methyl]cyclohexanone hydrochloride

  • MDL number MFCD00102580
  • Cas Number : 42036-65-7
  • Molecular Weight : 191.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM09904
Availability details for
Quantity
  • Availability
  • Add to Cart