ATC311763818 Atlantic Research Chemicals Ltd

9H-Carbazol-3ylboronic acid

  • MDL number MFCD04113810
  • Cas Number : 851524-97-5
  • Molecular Weight : 211.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=BA10001
Availability details for
Quantity
  • Availability
  • Add to Cart