ATC311763819 Atlantic Research Chemicals Ltd

2-Hydroxy-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene

  • MDL number MFCD12827861
  • Cas Number : 1260151-69-6
  • Molecular Weight : 270.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=BA10002
Availability details for
Quantity
  • Availability
  • Add to Cart