ATC311763835 Atlantic Research Chemicals Ltd

2,3-Diaminobenzoic acid

  • MDL number MFCD00137818
  • Cas Number : 603-81-6
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001102
Availability details for
Quantity
  • Availability
  • Add to Cart