ATC311763846 Atlantic Research Chemicals Ltd

4-Amino-3-iodobenzoic acid

  • MDL number MFCD01660550
  • Cas Number : 2122-63-6
  • Molecular Weight : 263.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001112
Availability details for
Quantity
  • Availability
  • Add to Cart