ATC311763866 Atlantic Research Chemicals Ltd

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD03011525
  • Cas Number : 81282-82-8
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001132
Availability details for
Quantity
  • Availability
  • Add to Cart