ATC311763911 Atlantic Research Chemicals Ltd

(4-Morpholin-4-ylphenyl)acetic acid

  • MDL number MFCD01995085
  • Cas Number : 26577-57-1
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001174
Availability details for
Quantity
  • Availability
  • Add to Cart