ATC311763914 Atlantic Research Chemicals Ltd

2,3,5,6-Tetrachloroterephthalic acid

  • MDL number MFCD00013974
  • Cas Number : 2136-79-0
  • Molecular Weight : 303.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001177
Availability details for
Quantity
  • Availability
  • Add to Cart