ATC311763924 Atlantic Research Chemicals Ltd

2-(2,6-Difluorophenyl)-2-methylpropanoic acid

  • MDL number MFCD11036902
  • Cas Number : 1216838-87-7
  • Molecular Weight : 200.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001186
Availability details for
Quantity
  • Availability
  • Add to Cart