ATC311763977 Atlantic Research Chemicals Ltd

3-(3,5-Dibromo-4-hydroxyphenyl)propanoic acid

  • MDL number MFCD00816760
  • Cas Number : 13811-12-6
  • Molecular Weight : 323.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001238
Availability details for
Quantity
  • Availability
  • Add to Cart