ATC311764005 Atlantic Research Chemicals Ltd

3-Amino-2,6-difluorobenzoic acid

  • MDL number MFCD07368833
  • Cas Number : 83141-11-1
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001266
Availability details for
Quantity
  • Availability
  • Add to Cart