ATC311764026 Atlantic Research Chemicals Ltd

(2S)-2-Benzyl-3-[(tert-butoxycarbonyl)amino]propanoic acid

  • MDL number MFCD07372495
  • Cas Number : 262301-38-2
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001286
Availability details for
Quantity
  • Availability
  • Add to Cart