ATC311764037 Atlantic Research Chemicals Ltd

2-(Aminosulphonyl)benzoic acid

  • MDL number MFCD00127704
  • Cas Number : 632-24-6
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001297
Availability details for
Quantity
  • Availability
  • Add to Cart