ATC311764077 Atlantic Research Chemicals Ltd

(2,4-Dimethylphenoxy)acetic acid

  • MDL number MFCD00014355
  • Cas Number : 13334-49-1
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001335
Availability details for
Quantity
  • Availability
  • Add to Cart