ATC311764117 Atlantic Research Chemicals Ltd

3,4-Dimethyl-2-iodobenzoic acid

  • MDL number MFCD14666470
  • Cas Number : 129833-31-4
  • Molecular Weight : 276.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001375
Availability details for
Quantity
  • Availability
  • Add to Cart