ATC311764127 Atlantic Research Chemicals Ltd

3-(Morpholin-4-yl)benzoic acid

  • MDL number MFCD06659078
  • Cas Number : 215309-00-5
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001385
Availability details for
Quantity
  • Availability
  • Add to Cart