ATC311764138 Atlantic Research Chemicals Ltd

(4-Nitrophenoxy)acetic acid

  • MDL number MFCD00017030
  • Cas Number : 1798-11-4
  • Molecular Weight : 197.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001396
Availability details for
Quantity
  • Availability
  • Add to Cart