ATC311764144 Atlantic Research Chemicals Ltd

2-Iodo-4-methoxybenzoic acid

  • MDL number MFCD09909619
  • Molecular Weight : 278.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001401
Availability details for
Quantity
  • Availability
  • Add to Cart