ATC311764151 Atlantic Research Chemicals Ltd

(2,3,5-Trifluorophenyl)acetic acid

  • MDL number MFCD00083547
  • Cas Number : 132992-28-0
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001408
Availability details for
Quantity
  • Availability
  • Add to Cart