ATC311764188 Atlantic Research Chemicals Ltd

2-Amino-5,6-difluorobenzoic acid

  • MDL number MFCD06808540
  • Cas Number : 442134-72-7
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001445
Availability details for
Quantity
  • Availability
  • Add to Cart