ATC311764192 Atlantic Research Chemicals Ltd

2,6-Dibromobenzoic acid

  • MDL number MFCD00234254
  • Cas Number : 601-84-3
  • Molecular Weight : 279.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001449
Availability details for
Quantity
  • Availability
  • Add to Cart