ATC311764230 Atlantic Research Chemicals Ltd

4-Fluoro-2,6-dimethylbenzoic acid

  • MDL number MFCD06208279
  • Cas Number : 16633-50-4
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00172
Availability details for
Quantity
  • Availability
  • Add to Cart