ATC311764275 Atlantic Research Chemicals Ltd

5-Phenylnicotinic Acid

  • MDL number MFCD03086176
  • Cas Number : 10177-12-5
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00211
Availability details for
Quantity
  • Availability
  • Add to Cart