ATC311764290 Atlantic Research Chemicals Ltd

6-(1H-Imidazol-1-yl)nicotinic acid

  • MDL number MFCD00140821
  • Cas Number : 216955-75-8
  • Molecular Weight : 189.173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA002123
Availability details for
Quantity
  • Availability
  • Add to Cart