ATC311764302 Atlantic Research Chemicals Ltd

2-(Methylamino)isonicotinic acid

  • MDL number MFCD07643263
  • Cas Number : 876717-53-2
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA002134
Availability details for
Quantity
  • Availability
  • Add to Cart