ATC311764318 Atlantic Research Chemicals Ltd

5-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid

  • MDL number MFCD09991768
  • Cas Number : 848308-47-4
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA002150
Availability details for
Quantity
  • Availability
  • Add to Cart