ATC311764324 Atlantic Research Chemicals Ltd

5-Nitropyridine-2-carboxylic acid

  • MDL number MFCD04114182
  • Cas Number : 30651-24-2
  • Molecular Weight : 168.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA002156
Availability details for
Quantity
  • Availability
  • Add to Cart