ATC311764330 Atlantic Research Chemicals Ltd

6-(Benzyloxy)pyridine-2-carboxylic acid

  • MDL number MFCD11100039
  • Cas Number : 149744-21-8
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA002162
Availability details for
Quantity
  • Availability
  • Add to Cart