ATC311764339 Atlantic Research Chemicals Ltd

2,2'-Bipyridine-6-carboxylic acid

  • MDL number MFCD11100046
  • Cas Number : 4392-87-4
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA002170
Availability details for
Quantity
  • Availability
  • Add to Cart