ATC311764528 Atlantic Research Chemicals Ltd

5-Chloro-2-methylnicotinic acid

  • MDL number MFCD11557228
  • Cas Number : 1092286-30-0
  • Molecular Weight : 171.582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA002348
Availability details for
Quantity
  • Availability
  • Add to Cart