ATC311764566 Atlantic Research Chemicals Ltd

6-Chloro-3-(trifluoromethyl)pyridine-2-carboxylic acid

  • MDL number MFCD08235206
  • Cas Number : 796090-24-9
  • Molecular Weight : 225.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00262
Availability details for
Quantity
  • Availability
  • Add to Cart