ATC311764581 Atlantic Research Chemicals Ltd

5-Cyano-6-methylnicotinic acid

  • MDL number MFCD11505881
  • Cas Number : 1216866-96-4
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00278
Availability details for
Quantity
  • Availability
  • Add to Cart