ATC311764603 Atlantic Research Chemicals Ltd

2-Aminothiophene-3-carboxylic Acid

  • MDL number MFCD03426102
  • Cas Number : 56387-08-7
  • Molecular Weight : 143.165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA00306
Availability details for
Quantity
  • Availability
  • Add to Cart