ATC311764612 Atlantic Research Chemicals Ltd

3-Aminothiophene-2-carboxylic acid

  • MDL number MFCD03426100
  • Cas Number : 55341-87-2
  • Molecular Weight : 143.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00326
Availability details for
Quantity
  • Availability
  • Add to Cart