ATC311764631 Atlantic Research Chemicals Ltd

5-Chloro-4-methoxythiophene-3-carboxylic acid

  • MDL number MFCD00067999
  • Cas Number : 133659-14-0
  • Molecular Weight : 192.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00345
Availability details for
Quantity
  • Availability
  • Add to Cart