ATC311764654 Atlantic Research Chemicals Ltd

2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid

  • MDL number MFCD02677730
  • Cas Number : 117621-06-4
  • Molecular Weight : 236.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00420
Availability details for
Quantity
  • Availability
  • Add to Cart