ATC311764912 Atlantic Research Chemicals Ltd

5-Bromo-1-benzothiophene-3-carboxylic acid

  • MDL number MFCD06797473
  • Cas Number : 7312-24-5
  • Molecular Weight : 257.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA01705
Availability details for
Quantity
  • Availability
  • Add to Cart