ATC311764924 Atlantic Research Chemicals Ltd

1H-Indole-2-acetic acid (as Na salt)

  • MDL number MFCD00957069
  • Cas Number : 32588-36-6
  • Molecular Weight : 175.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA01904
Availability details for
Quantity
  • Availability
  • Add to Cart