ATC311764928 Atlantic Research Chemicals Ltd

2-Chloro-1H-indole-3-carboxylic acid

  • MDL number MFCD01719046
  • Cas Number : 54778-20-0
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA01908
Availability details for
Quantity
  • Availability
  • Add to Cart