ATC311764943 Atlantic Research Chemicals Ltd

2-Methyl-1,3-benzoxazole-5-carboxylic acid

  • MDL number MFCD09759125
  • Cas Number : 90322-32-0
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA02102
Availability details for
Quantity
  • Availability
  • Add to Cart