ATC311764962 Atlantic Research Chemicals Ltd

3-(Trifluoromethyl)quinoline-4-carboxylic acid

  • MDL number MFCD08458858
  • Cas Number : 588702-65-2
  • Molecular Weight : 241.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA02609
Availability details for
Quantity
  • Availability
  • Add to Cart