ATC311765080 Atlantic Research Chemicals Ltd

4-Benzylmorpholine-2-carboxylic acid

  • MDL number MFCD06410628
  • Cas Number : 769087-80-1
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA04106
Availability details for
Quantity
  • Availability
  • Add to Cart