ATC311765088 Atlantic Research Chemicals Ltd

1-[(Benzyloxy)carbonyl]pyrrolidine-2-carboxylic acid

  • MDL number MFCD00063229
  • Cas Number : 5618-96-2
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA04501
Availability details for
Quantity
  • Availability
  • Add to Cart