ATC311765112 Atlantic Research Chemicals Ltd

1-Methylpyrrolidine-3-carboxylic acid

  • MDL number MFCD09055221
  • Cas Number : 25712-60-1
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA04525
Availability details for
Quantity
  • Availability
  • Add to Cart