ATC311765162 Atlantic Research Chemicals Ltd

5-Oxo-1-(2-phenylethyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD00973748
  • Cas Number : 30380-70-2
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA04576
Availability details for
Quantity
  • Availability
  • Add to Cart