ATC311765176 Atlantic Research Chemicals Ltd

1-Methyl-1H-1,2,4-triazole-5-carboxylic acid

  • MDL number MFCD11100092
  • Cas Number : 815588-93-3
  • Molecular Weight : 127.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA04703
Availability details for
Quantity
  • Availability
  • Add to Cart