ATC311765289 Atlantic Research Chemicals Ltd

Ceftriaxone Sodium Salt

  • MDL number MFCD09749665
  • Cas Number : 74578-69-1
  • Molecular Weight : 564.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA100108
Availability details for
Quantity
  • Availability
  • Add to Cart