ATC311765291 Atlantic Research Chemicals Ltd

2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid

  • MDL number MFCD00244615
  • Cas Number : 21801-79-6
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA10011
Availability details for
Quantity
  • Availability
  • Add to Cart