ATC311765292 Atlantic Research Chemicals Ltd

5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid

  • MDL number MFCD01464030
  • Cas Number : 4242-18-6
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA100110
Availability details for
Quantity
  • Availability
  • Add to Cart